Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Bromide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
7647-15-6
BrNa
102.894
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a
253881
CHEBI:63004
sodium;bromide
[Na+].[Br-]
7647-15-6
BrNa
102.894
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a
253881
CHEBI:63004
sodium;bromide
[Na+].[Br-]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 102.89
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 3.21kg
Quantity 2.5kg
Chemical Name or Material Sodium Bromide