Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate, 99.8%, for analysis, anhydrous, powder, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic bottle
497-19-8
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
497-19-8
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
Adsorbent, Biological buffer
Boiling Point 1600.0°C
Melting Point 851.0°C
Packaging Plastic bottle
Loss on Drying 0.5% max. (300°C)
Color White
Quantity 1kg
Assay Percent Range 99.8%
Linear Formula Na2CO3
Fieser 07,332
Merck Index 15, 8731
Solubility Information Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Formula Weight 105.99
Physical Form Crystalline Powder or Granules
Percent Purity 99.8%
Grade Analysis
Chemical Name or Material Sodium carbonate, Anhydrous
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning