Alfa Aesar™ Sodium carbonate, Acculute Standard Volumetric Solution, Final Concentration 0.1N

Manufacturer: Alfa Aesar™ 88617LS

Quantity 6 units

 View more versions of this product

Catalog No. AA88617LS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 497-19-8
    Molecular Formula Na2CO3
    Formula Weight 105.99
    MDL Number MFCD00003494
    Chemical Name or Material Sodium carbonate Acculute Standard Volumetric Solution, Final Concentration 0.1N

    Specifications

    Formula Weight 105.99
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Quantity 6 units
    Molecular Formula Na2CO3
    MDL Number MFCD00003494
    CAS 497-19-8
    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Liquid
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Chemical Name or Material Sodium carbonate Acculute Standard Volumetric Solution, Final Concentration 0.1N

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit