Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate, 99.95%, extra pure, anhydrous, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Plastic bottle
497-19-8
CNa2O3
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
497-19-8
CNa2O3
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
Adsorbent
Aluminum (Al) 10ppm max.
Assay Percent Range 99.95%
Linear Formula Na2CO3
Fieser 07,332
Merck Index 15,8731
Solubility Information Solubility in water: 22g/100mL (20°C). Other solubilities: soluble in glycerol,insoluble in alcohol
Formula Weight 105.99
Physical Form Fine Crystalline Powder
Percent Purity 99.95%
Grade Extra Pure
Calcium (Ca) 50ppm max.
Chloride 20ppm max
Chlorine (Cl) 20ppm max.
Heavy Metals (as Pb) 5ppm max.
Insoluble in Water 0.01% max.
Insoluble Matter 0.01% max. (in water)
Iron (Fe) 5ppm max.
Loss on Drying 1% (285°C) max.
Packaging Plastic bottle
Phosphate 10ppm max.
Potassium (K) 100ppm max.
Silicon Dioxide 50ppm max.
Solubility Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Total Nitrogen Compounds 0.001% (as N) max.
Total Sulfur 0.005% max.
Boiling Point 1600.0°C
Color White
Melting Point 851.0°C
Quantity 500g
Chemical Name or Material Sodium carbonate
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get med

GHS Signal Word: Warning

EINECSNumber : 207-838-8