Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, Anhydrous, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle
497-19-8
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
497-19-8
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 105.99
Physical Form Granular
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.53kg
Quantity 500g
Chemical Name or Material Sodium Carbonate, Anhydrous