Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, Anhydrous, Granular, N.F. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
3kg
500g
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle

Chemical Identifiers

497-19-8
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
CAS 497-19-8
Molecular Formula CNa2O3
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES C(=O)([O-])[O-].[Na+].[Na+]
IUPAC Name disodium;carbonate
Molecular Weight (g/mol) 105.988
PubChem CID 10340
ChEBI CHEBI:29377
Formula Weight 105.99
Physical Form Granular
Packaging Poly Pail
Specific Gravity 1L = 2.53kg
Quantity 3kg
Chemical Name or Material Sodium Carbonate, Anhydrous
Grade FCC/NF
Show More Show Less