Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, Anhydrous, Granular, N.F. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
500g
3kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle

Chemical Identifiers

497-19-8
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
SMILES C(=O)([O-])[O-].[Na+].[Na+]
IUPAC Name disodium;carbonate
Formula Weight 105.99
Physical Form Granular
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.53kg
Quantity 500g
Chemical Name or Material Sodium Carbonate, Anhydrous
Grade FCC/NF