Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, Anhydrous, NF, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
497-19-8
CNa2O3
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium carbonate
[Na+].[Na+].[O-]C([O-])=O
497-19-8
CNa2O3
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium carbonate
[Na+].[Na+].[O-]C([O-])=O

SO147, 497-19-8, Na2CO3

Spectrum™ Sodium Carbonate, Anhydrous, NF is a pH regulator excipient. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade NF
CAS Max % 1
Percent Purity 99.5 to 100.5%
Packaging Poly Bottle
Water 0.005
Quantity 2.5 kg