Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, Anhydrous, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle

Chemical Identifiers

497-19-8
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 105.99
Physical Form Powder
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.53kg
Quantity 500g
Chemical Name or Material Sodium Carbonate, Anhydrous
Grade Baker Analyzed™ ACS Reagent