Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, Anhydrous, ULTREX™ Ultrapure Reagent, J.T.Baker™

Click to view available options
Quantity:
25g
100g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

497-19-8
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
Formula Weight 105.99
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.53kg
Quantity 25g
Chemical Name or Material Sodium Carbonate, Anhydrous
Grade ULTREX™ Ultrapure Reagent