Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

5968-11-6
CH2Na2O4
124.00
MQRJBSHKWOFOGF-UHFFFAOYSA-L
2735133

High quality chemicals for laboratory and specialized industrial use

CAS 5968-11-6
Molecular Formula CH2Na2O4
InChI Key MQRJBSHKWOFOGF-UHFFFAOYSA-L
SMILES C(=O)([O-])[O-].O.[Na+].[Na+]
IUPAC Name disodium;carbonate;hydrate
Molecular Weight (g/mol) 124.00
PubChem CID 2735133
Formula Weight 124
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.25kg
Quantity 2.5kg
Chemical Name or Material Sodium Carbonate, Monohydrate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less