Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, Monohydrate, Crystal, N.F. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
5968-11-6
CH2Na2O4
124.00
MQRJBSHKWOFOGF-UHFFFAOYSA-L
2735133
disodium;carbonate;hydrate
C(=O)([O-])[O-].O.[Na+].[Na+]
5968-11-6
CH2Na2O4
124.00
MQRJBSHKWOFOGF-UHFFFAOYSA-L
2735133
disodium;carbonate;hydrate
C(=O)([O-])[O-].O.[Na+].[Na+]
Formula Weight 124
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.25kg
Quantity 500g
Grade FCC/NF
Chemical Name or Material Sodium Carbonate, Monohydrate