Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, Monohydrate, Crystal, N.F. - F.C.C., J.T. Baker™

Catalog No. 02004013 Shop All Avantor Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 5968-11-6
Molecular Formula CH2Na2O4
Molecular Weight (g/mol) 124.00
InChI Key MQRJBSHKWOFOGF-UHFFFAOYSA-L
Synonym sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
PubChem CID 2735133
IUPAC Name disodium;carbonate;hydrate
SMILES C(=O)([O-])[O-].O.[Na+].[Na+]

Specifications

Physical Form Crystals
Quantity 2.5 kg
Formula Weight 124
Grade FCC/NF
Packaging Poly Bottle
Specific Gravity 1L = 2.25kg
Chemical Name or Material Sodium Carbonate, Monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.