Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate Monohydrate (Granular/NF/FCC/EP/BP), Fisher Chemical

Click to view available options
Quantity:
2.5kg
Packaging:
Poly Bottle
5968-11-6
CH2Na2O4
124.00
MFCD00149177
MQRJBSHKWOFOGF-UHFFFAOYSA-L
sodium carbonate monohydrate,disodium carbonate monohydrate,sodium carbonate, monohydrate,carbonic acid disodium salt, monohydrate,sodium carbonate hydrate,carbonic acid sodium salt monohydrate,disodium carbonate hydrate,disodium hydrate carbonate,na2co3 water,acmc-20ajop
2735133
disodium;carbonate;hydrate
C(=O)([O-])[O-].O.[Na+].[Na+]
5968-11-6
CH2Na2O4
124.00
MFCD00149177
MQRJBSHKWOFOGF-UHFFFAOYSA-L
sodium carbonate monohydrate,disodium carbonate monohydrate,sodium carbonate, monohydrate,carbonic acid disodium salt, monohydrate,sodium carbonate hydrate,carbonic acid sodium salt monohydrate,disodium carbonate hydrate,disodium hydrate carbonate,na2co3 water,acmc-20ajop
2735133
disodium;carbonate;hydrate
C(=O)([O-])[O-].O.[Na+].[Na+]
Alkali Hydroxide and Bicarbonates Pass Test (BP),Pass Test (EP),Pass Test (EP/BP)
Appearance of Solution Pass Test (BP),Pass Test (EP),Pass Test (EP/BP)
Arsenic (As) 5ppm max. (BP),5ppm max. (EP),5ppm max. (EP/BP)
Bacterial Endotoxins Report EU/mg (EP)
Formula Weight 124.01
Physical Form Powder/Solid
Percent Purity 83.0 to 87.5% (EP)
Grade BP/EP/FCC/NF
Chloride 125ppm max. (BP),125ppm max. (EP),125ppm max. (EP/BP)
Density 2.25g/cm³
Endotoxin Report EU/mg (NF)
Heavy Metals 50ppm max. (BP),50ppm max. (EP),50ppm max. (EP/BP)
Heavy Metals (as Pb) 0.001% max. (NF),10mg/kg max. (FCC),10mg/kg max. (FCC), 0.001% max. (NF)
Identification Pass Test (BP),Pass Test (EP),Pass Test (FCC),Pass Test (NF),Pass Test (NF/FCC/EP/BP)
Iron (Fe) 50ppm max. (BP),50ppm max. (EP),50ppm max. (EP/BP)
Lead (Pb) 4mg/kg max. (FCC),4mg/kg max. (FCC/NF),4mg/kg max. (NF)
Loss on Drying 12.0 to 15.0% (FCC)
Packaging Poly Bottle
Residual Solvents Meets Requirements (BP),Meets Requirements (EP),Meets Requirements (NF),Meets Requirements (NF/EP/BP)
Sulfate 250ppm max. (BP),250ppm max. (EP),250ppm max. (EP/BP)
Water 12 to 15% (NF)
Color White
Melting Point 851°C
Quantity 2.5kg
Chemical Name or Material Sodium Carbonate Monohydrate
Show More Show Less