Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Poly Bottle
497-19-8,7732-18-5
CNa2O3
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium carbonate
[Na+].[Na+].[O-]C([O-])=O
497-19-8,7732-18-5
CNa2O3
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium carbonate
[Na+].[Na+].[O-]C([O-])=O

S-148, 497-19-8

Spectrum™ Sodium Carbonate TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

CAS Max % 0.001,0.999
Grade Chemical Solution
Packaging Poly Bottle
Quantity 100 mL