Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate TS, (U.S.P. Test Solution), Spectrum™ Chemical
SDP

Catalog Number 18606372
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Poly Bottle
497-19-8, 7732-18-5
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium carbonate
[Na+].[Na+].[O-]C([O-])=O
497-19-8, 7732-18-5
CNa2O3
105.99
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium carbonate
[Na+].[Na+].[O-]C([O-])=O

S-148, 497-19-8

Spectrum™ Chemical Sodium Carbonate TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

CAS Max % 0.001, 0.999
Grade Chemical Solution
Packaging Poly Bottle
Quantity 1 L
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit