Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Cetostearyl Sulfate, NF, EP, Spectrum™

Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
68955-20-4
C17H35NaO4S
358.51
HHURSJAUVYNJBT-UHFFFAOYSA-M
sodium heptadecyl sulfate
[Na+].CCCCCCCCCCCCCCCCCOS([O-])(=O)=O
68955-20-4
C17H35NaO4S
358.51
HHURSJAUVYNJBT-UHFFFAOYSA-M
sodium heptadecyl sulfate
[Na+].CCCCCCCCCCCCCCCCCOS([O-])(=O)=O

S1980, 68955-20-4

Spectrum™ Sodium Cetostearyl Sulfate, NF, EP
Packaging Poly Bottle
Quantity 100 g
Water 0.015
CAS Max % 1