Sodium chlorate, ACS, 99.0% min, Alfa Aesar™

Maximum level of impurities: Insoluble matter 0.005%, BrO3 0.015%, Cl 0.005%, SO4 0.001%, Heavy metals (as Pb) 0.001%, Fe 5ppm, Ca 0.002%, Mg 0.002%, K 0.01%

Manufacturer: Alfa Aesar 4440836

Quantity 500g

 View more versions of this product

Catalog No. AA4440836

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7775-09-9
    Physical Form Crystalline
    Assay Percent Range 99% min.
    Molecular Formula NaClO3
    Formula Weight 106.45
    MDL Number MFCD00003479
    Chemical Name or Material Sodium chlorate, For analysis ACS
    Grade ACS Grade

    Specifications

    Physical Form Crystalline
    Formula Weight 106.45
    Quantity 500g
    Color Colorless
    DOT Information DOT Class: 5.1, Packaging Group: II
    Density 2.5g/cm3 at 20°C
    EINECS Number 231-887-4
    Odor Odorless
    Assay Percent Range 99% min.
    Molecular Formula NaClO3
    MDL Number MFCD00003479
    CAS 7775-09-9
    Health Hazard 1 H271-H302-H411-H401: May cause fire or explosion; strong oxidizer. Harmful if swallowed. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.
    Chemical Name or Material Sodium chlorate, For analysis ACS
    Grade ACS Grade

    H271-H302-H411-H401: May cause fire or explosion; strong oxidizer. Harmful if swallowed. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit