Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Chloride, 0.9% (w/v), Isotonic Saline, Ricca Chemical
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Concentration or Composition (by Analyte or Components) | 0.9% (w/v) |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous |
Quantity | 500 mL |
Sterility | Non-sterile |
Chemical Name or Material | Sodium Chloride, Isotonic Saline |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction