Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Chloride, 9% (w/v), Ricca Chemical
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Color | Colorless |
CAS Min % | 90.0, 8.1 |
CAS Max % | 94.0, 8.43 |
Physical Form | Liquid |
Quantity | 10 L |
Concentration or Composition (by Analyte or Components) | 9% (w/v) Aqueous Solution |
Packaging | Cubitainer |
Chemical Name or Material | Sodium Chloride |
Safety and Handling
ShelfLife | 24 months |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction