Sodium chloride, ACS, 99.0% min., Alfa Aesar™

Manufacturer: Alfa Aesar 12314A3

Quantity 2kg

 View more versions of this product

Catalog No. AA12314A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7647-14-5
    Physical Form Crystalline
    Assay Percent Range 99.0% min
    Molecular Formula NaCl
    Formula Weight 58.44g/mol
    MDL Number MFCD00003477
    Chemical Name or Material Sodium chloride, For analysis ACS

    Specifications

    Density 2.165g/mL
    Quantity 2kg
    CAS 7647-14-5
    Physical Form Crystalline
    Assay Percent Range 99.0% min
    Molecular Formula NaCl
    Formula Weight 58.44g/mol
    MDL Number MFCD00003477
    EINECS Number 231-598-3
    Chemical Name or Material Sodium chloride, For analysis ACS

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit