Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
1kg
Packaging:
Poly Bottle
7647-14-5
ClNa
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
5234
CHEBI:26710
sodium;chloride
[Na+].[Cl-]
7647-14-5
ClNa
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
5234
CHEBI:26710
sodium;chloride
[Na+].[Cl-]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 58.44
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.16kg
Quantity 500g
Chemical Name or Material Sodium Chloride