Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
1kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7647-14-5
ClNa
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
CHEBI:26710

High quality chemicals for laboratory and specialized industrial use

CAS 7647-14-5
Molecular Formula ClNa
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium;chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Formula Weight 58.44
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.16kg
Quantity 1kg
Chemical Name or Material Sodium Chloride
Grade Baker Analyzed™ ACS Reagent
Show More Show Less