Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Crystal, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
500g
1kg
2.5kg
5kg
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

7647-14-5
ClNa
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
CHEBI:26710
sodium;chloride
[Na+].[Cl-]

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography.

    For Liquid Chromatography and Molecular Biology applications
Formula Weight 58.44
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.16kg
Quantity 2.5kg
Chemical Name or Material Sodium Chloride
Grade Ultrapure Bioreagent