Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Crystal, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
1kg
2.5kg
500g
5kg
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

7647-14-5
ClNa
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
CHEBI:26710

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography.

    For Liquid Chromatography and Molecular Biology applications
CAS 7647-14-5
Molecular Formula ClNa
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium;chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Formula Weight 58.44
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.16kg
Quantity 500g
Chemical Name or Material Sodium Chloride
Grade Ultrapure Bioreagent
Show More Show Less