Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STE Buffer, Sodium Chloride-Tris-EDTA, 10X Solution, pH 8.0, Fisher BioReagents

Chemical Identifiers
CAS | 17372-87-1 |
---|---|
Molecular Formula | C20H12Br4Na2O8 |
Molecular Weight (g/mol) | 745.904 |
InChI Key | MASXMTNVNASWNH-UHFFFAOYSA-L |
PubChem CID | 91886399 |
IUPAC Name | disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate |
SMILES | C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+] |
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
BP24791
|
1L | Nalgene Poly Bottle |
Each for $82.00
Case of 6 EA for $392.70
Order in Bulk
Request bulk or custom formats |
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
BP2479500
|
500mL | Poly Bottle |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
Description
STE buffer is suitable for biomolecular lab procedures.Chemical Identifiers
17372-87-1 | |
745.904 | |
91886399 | |
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+] |
C20H12Br4Na2O8 | |
MASXMTNVNASWNH-UHFFFAOYSA-L | |
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate |
Specifications
Sodium Chloride-Tris-EDTA (STE) Buffer | |
7647-14-5, 60-00-4, 7647-01-0, 7732-18-5 | |
STE, Saline Tris–EDTA, TEN | |
Protease free | |
100°C | |
Undesignated |
10X, pH 8.0 | |
DNase free | |
Liquid | |
Filtered through a 5-micron filter. | |
8 |
Safety and Handling

Recommended Storage : RT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title