Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

STE Buffer, Sodium Chloride-Tris-EDTA, 1X Solution, pH 8.0, Fisher BioReagents
Description
STE buffer is suitable for biomolecular lab procedures.Specifications
Sodium Chloride-Tris-EDTA Buffer | |
7647-14-5, 60-00-4, 7647-01-0, 7732-18-5 | |
100°C | |
Liquid | |
100mL | |
Filtered through a 5-micron filter. | |
Protease free |
1X, pH 8.0 | |
Saline Tris–EDTA, STE | |
Undesignated | |
8 | |
DNase free | |
Poly Bottle |
Safety and Handling

Recommended Storage : RT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title