Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

STE Buffer, Sodium Chloride-Tris-EDTA, 1X Solution, pH 8.0, Fisher BioReagents
Description
- Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X STE buffer solution
- Eliminates the hassle of dilution or waiting for powder to dissolve
- 1X solution contains 10mM tris, 1mM EDTA, and 100mM NaCl
- Filtered through a 5 micron filter and autoclaved
- Tested for the absence of DNase, RNase, and protease
- STE buffer is used for DNA extraction protocols
- Intended for laboratory research use only
Specifications
Sodium Chloride-Tris-EDTA Buffer | |
7647-14-5, 60-00-4, 7647-01-0, 7732-18-5 | |
100°C | |
Liquid | |
1L | |
Filtered through a 5-micron filter. | |
Protease free |
1X, pH 8.0 | |
Saline Tris–EDTA, STE, TEN | |
Undesignated | |
8 | |
DNase free | |
Nalgene Poly Bottle |
Safety and Handling

Recommended Storage : RT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title