Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STE Buffer, Sodium Chloride-Tris-EDTA, 1X Solution, pH 8.0, Fisher BioReagents

Specifications
Chemical Name or Material | Sodium Chloride-Tris-EDTA Buffer |
---|---|
Name Note | 1X, pH 8.0 |
CAS | 7647-14-5, 60-00-4, 7647-01-0, 7732-18-5 |
Synonym | STE, Saline Tris–EDTA |
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
BP2478100
|
100mL | Poly Bottle |
Each for $17.25
Case of 6 EA for $107.77
Order in Bulk
Request bulk or custom formats |
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
BP24781
|
1L | Nalgene Poly Bottle |
Each for $76.00
Case of 6 EA for $363.39
Order in Bulk
Request bulk or custom formats |
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
BP2478500
|
500mL | Poly Bottle |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
Description
STE buffer is suitable for biomolecular lab procedures.Specifications
Sodium Chloride-Tris-EDTA Buffer | |
1X, pH 8.0 | |
7647-14-5, 60-00-4, 7647-01-0, 7732-18-5 | |
STE, Saline Tris–EDTA |
Safety and Handling

Recommended Storage : RT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title