Learn More
Sodium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media, a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | 1413°C |
Melting Point | 801°C |
Color | White |
pH | 6 to 9 |
Physical Form | Beads |
Quantity | 5 g |
Merck Index | 14,8599 |
Solubility Information | Soluble in water (358mg/ml at 20°C),alcohol (very slightly),glycerol (100mg/ml),and ammonia. Insoluble in hydrochloric acid. |
IUPAC Name | sodium;chloride |
Formula Weight | 58.44 |
Show More |
Safety and Handling
EINECSNumber | 231-598-3 |
RTECSNumber | VZ4725000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.