Alfa Aesar™ Sodium chloride, ultra dry, 99.998% (metals basis)

Manufacturer: Alfa Aesar™ 1368314

Quantity 25g

 View more versions of this product

Catalog No. AA1368314

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7647-14-5
    Formula Weight 58.44
    MDL Number MFCD00003477
    Merck Index 14,8599
    Chemical Name or Material Sodium chloride, Ultra dry

    Specifications

    Formula Weight 58.44
    Packaging Ampule
    Odor Odorless
    CAS 7647-14-5
    Merck Index 14,8599
    Boiling Point 1413°C
    Melting Point 801°C
    pH 6 to 9
    Density 2.165g/mL
    Mesh Size −10 Mesh
    Physical Form Beads
    MDL Number MFCD00003477
    Color White
    TSCA TSCA
    Refractive Index 1.5442
    Sensitivity Hygroscopic
    Storage Note 1
    • Ampouled under argon
    Quantity 25g
    RTECS Number VZ4725000
    Chemical Name or Material Sodium chloride, Ultra dry
    Assay 99.998%, (metals basis)

    • Ampouled under argon

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit