Alfa Aesar™ Sodium chloroacetate, 98%

Manufacturer: Alfa Aesar™ A123790C

Quantity 10,000g

 View more versions of this product

Catalog No. AAA123790C

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3926-62-3
    Molecular Formula C2H2ClNaO2
    Linear Formula ClCH2CO2Na
    Formula Weight 116.48
    MDL Number MFCD00002684
    UN Number UN2659
    Beilstein 3597157
    Merck Index 14,2112
    Synonym Chloroacetic acid sodium salt
    Chemical Name or Material Sodium chloroacetate

    Specifications

    Formula Weight 116.48
    Physical Form Powder
    Assay Percent Range 98%
    Storage Note 1 Hygroscopic
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P362-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Molecular Formula C2H2ClNaO2
    Linear Formula ClCH2CO2Na
    CAS 3926-62-3
    Beilstein 3597157
    Merck Index 14,2112
    Melting Point ≈199°C (dec.)
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SODIUM CHLOROACETATE
    MDL Number MFCD00002684
    UN Number UN2659
    Flash Point 270°C (518°F)
    RTECS Number AG1400000
    TSCA TSCA
    Synonym Chloroacetic acid sodium salt
    Health Hazard 2 GHS H Statement
    H301-H315
    Toxic if swallowed.
    Causes skin irritation.
    pH 6
    Odor Characteristic
    Quantity 10,000g
    Chemical Name or Material Sodium chloroacetate
    Assay 98%

    Description

    Applications
    Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.

    Solubility
    Soluble in water, ether, chloroform, benzene and alcohol.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H301-H315
    Toxic if swallowed.
    Causes skin irritation.

    GHS P Statement
    P280-P301+P310-P362-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Hygroscopic

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit