Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium chloroacetate, 98%

Manufacturer:  Alfa Aesar™ A1237930

Quantity 250g

 View more versions of this product

Catalog No. AAA1237930


  Check Availability
Add to cart

Specifications

Description

Applications
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.

Solubility
Soluble in water, ether, chloroform, benzene and alcohol.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

3926-62-3
ClCH2CO2Na
MFCD00002684
3597157
Chloroacetic acid sodium salt
C2H2ClNaO2
116.48
UN2659
14,2112
Sodium chloroacetate

Specifications

3926-62-3
98%
ClCH2CO2Na
MFCD00002684
3597157
Chloroacetic acid sodium salt
≈199°C (dec.)
270°C (518°F)
Sodium chloroacetate
Powder
C2H2ClNaO2
116.48
UN2659
14,2112
6
250g
Characteristic
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315
Toxic if swallowed.
Causes skin irritation.

GHS P Statement
P280-P301+P310-P362-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SODIUM CHLOROACETATE

RTECSNumber : AG1400000

TSCA : TSCA

storageNote1 : Hygroscopic

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit