Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium chloroacetate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10,000 g
250 g
2500 g
500 g
Description
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.
Solubility
Soluble in water, ether, chloroform, benzene and alcohol.
Notes
Incompatible with strong oxidizing agents.
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.
Solubility
Soluble in water, ether, chloroform, benzene and alcohol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∼199°C (decomposition) |
pH | 6 |
Flash Point | 270°C (518°F) |
Odor | Characteristic |
Assay Percent Range | 98% |
Linear Formula | ClCH2CO2Na |
Quantity | 250 g |
UN Number | UN2659 |
Beilstein | 3597157 |
Merck Index | 14,2112 |
Show More |
Safety and Handling
Hazard Category | H301-H315 |
Hazard Statement | GHS H Statement H301-H315 Toxic if swallowed. Causes skin irritation. |
Precautionary Statement | P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SODIUM CHLOROACETATE |
EINECSNumber | 223-498-3 |
RTECSNumber | AG1400000 |
TSCA | Yes |
storageNote1 | Hygroscopic |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction