Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium chlorodifluoroacetate, 97%

Click to view available options
Quantity:
25g
100g
500g
1895-39-2
C2ClF2NaO2
152.457
MFCD00064771
MRTAVLDNYYEJHK-UHFFFAOYSA-M
sodium chlorodifluoroacetate,sodium 2-chloro-2,2-difluoroacetate,chlorodifluoroacetic acid sodium salt,sodium chloro difluoro acetate,acetic acid, chlorodifluoro-, sodium salt,sodiumchlorodifluoroacetate,difluorochloroacetic acid sodium salt,sodium difluorochloroacetate,chlorodifluoroacetic acid sodium
2734985
sodium;2-chloro-2,2-difluoroacetate
C(=O)(C(F)(F)Cl)[O-].[Na+]
1895-39-2
C2ClF2NaO2
152.457
MFCD00064771
MRTAVLDNYYEJHK-UHFFFAOYSA-M
sodium chlorodifluoroacetate,sodium 2-chloro-2,2-difluoroacetate,chlorodifluoroacetic acid sodium salt,sodium chloro difluoro acetate,acetic acid, chlorodifluoro-, sodium salt,sodiumchlorodifluoroacetate,difluorochloroacetic acid sodium salt,sodium difluorochloroacetate,chlorodifluoroacetic acid sodium
2734985
sodium;2-chloro-2,2-difluoroacetate
C(=O)(C(F)(F)Cl)[O-].[Na+]
Applications
It is used as pharmaceutical intermediate. Reactions of aldehydes with triphenylphosphine and sodium chlorodifluoroacetate yield 1,1-difluoro olefins. It is a general reaction and a one step synthesis.

Solubility
Soluble in dimethoxyethane, diglyme (0.60 g mL-1), triglyme, dimethyl formamide.

Notes
Store at room temperature. Keep away from oxidizing agents.
Quantity 500g
Melting Point ~195°C (decomposition)
Beilstein 4162363
Solubility Information Soluble in dimethoxyethane,diglyme (0.60 g mL-1),triglyme,dimethyl formamide.
Formula Weight 152.46
Percent Purity 97%
Chemical Name or Material Sodium chlorodifluoroacetate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-586-0

TSCA : Yes

Recommended Storage : Ambient temperatures