Alfa Aesar™ Sodium chlorodifluoroacetate

Manufacturer: Alfa Aesar™ 1676109

Quantity 10g

 View more versions of this product

Catalog No. AA1676109

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1895-39-2
    Formula Weight 152.46
    UN Number UN3288
    Chemical Name or Material Sodium chlorodifluoroacetate

    Specifications

    Formula Weight 152.46
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3288
    Quantity 10g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    TSCA TSCA
    Physical Form Crystalline
    CAS 1895-39-2
    Melting Point 196° to 198°C
    Chemical Name or Material Sodium chlorodifluoroacetate

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit