Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chromate, 4-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
10034-82-9
CrH8Na2O8
234.032
AVHUVTPOXZEIRG-UHFFFAOYSA-N
sodium chromate tetrahydrate,cro4.2na.4h2o,acmc-20ajol,disodium chromate tetrahydrate,ksc492q3d,sodium chromate tetrahydrate,disodium chromate 2-tetrahydrate,sodium chromate tetrahydrate, reagent grade,sodium chromate tetrahydrate, p.a
22146515
disodium;dioxido(dioxo)chromium;tetrahydrate
O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]
10034-82-9
CrH8Na2O8
234.032
AVHUVTPOXZEIRG-UHFFFAOYSA-N
sodium chromate tetrahydrate,cro4.2na.4h2o,acmc-20ajol,disodium chromate tetrahydrate,ksc492q3d,sodium chromate tetrahydrate,disodium chromate 2-tetrahydrate,sodium chromate tetrahydrate, reagent grade,sodium chromate tetrahydrate, p.a
22146515
disodium;dioxido(dioxo)chromium;tetrahydrate
O.O.O.O.[O-][Cr](=O)(=O)[O-].[Na+].[Na+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 234.03
Physical Form Crystals
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Quantity 500g
Chemical Name or Material Sodium Chromate, 4-Hydrate