Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chromate, 5% w/v, Reagents

Catalog No. 01333842
Change view
Click to view available options
Quantity:
1 L
4 L
Catalog No. Quantity
01-333-842 4 L
01-333-843 1 L
2 options

Catalog No. 01-333-842

Supplier: Reagents Holdings Llc CS1042004A

Only null left
Add to Cart
Add to Cart

Sodium Chromate, 5% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 10034-82-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Chromium disodium oxide
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Yellow
Physical Form Liquid
Quantity 4 L
Percent Purity 5%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.02
Chemical Name or Material Sodium Chromate, 5% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.