Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chromate, 98+%, anhydrous, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Plastic bottle
7775-11-3
CrNa2O4
161.97
MFCD00003484
PXLIDIMHPNPGMH-UHFFFAOYSA-N
sodium chlorate,chloric acid, sodium salt,asex,chlorate de sodium,chlorsaure,tumbleleaf,atlacide,desolet,kusatol,rasikal
516902
CHEBI:65242
sodium;chlorate
[Na+].[Na+].[O-][Cr]([O-])(=O)=O
7775-11-3
CrNa2O4
161.97
MFCD00003484
PXLIDIMHPNPGMH-UHFFFAOYSA-N
sodium chlorate,chloric acid, sodium salt,asex,chlorate de sodium,chlorsaure,tumbleleaf,atlacide,desolet,kusatol,rasikal
516902
CHEBI:65242
sodium;chlorate
[Na+].[Na+].[O-][Cr]([O-])(=O)=O
Oxidation reagent
Assay Percent Range 98% min. (Iodometry)
Linear Formula Na2CrO4
Merck Index 15, 8736
Solubility Information Solubility in water: 530g/L (20°C). Other solubilities: soluble in methanol,slightly soluble in ethanol
Formula Weight 161.97
Physical Form Crystals or Crystalline Powder
Percent Purity 98+%
Packaging Plastic bottle
Solubility Solubility in water: 530g/L (20°C). Other solubilities: soluble in methanol, slightly soluble in ethanol
Color Yellow
Melting Point 792.0°C
Quantity 25g
Chemical Name or Material Sodium chromate, Anhydrous
Show More Show Less

GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing diffi

GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

EINECSNumber : 231-889-5