Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chromate, 98+%, anhydrous, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Plastic bottle
7775-11-3
CrNa2O4
161.97
MFCD00003484
PXLIDIMHPNPGMH-UHFFFAOYSA-N
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
516902
CHEBI:65242
sodium;chlorate
[Na+].[Na+].[O-][Cr]([O-])(=O)=O
7775-11-3
CrNa2O4
161.97
MFCD00003484
PXLIDIMHPNPGMH-UHFFFAOYSA-N
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
516902
CHEBI:65242
sodium;chlorate
[Na+].[Na+].[O-][Cr]([O-])(=O)=O

Sodium chromate, 98+%, CrNa2O4, CAS Number-7775-11-3, asex, atlacide, chlorate de sodium, chloric acid, sodium salt, chlorsaure, desolet, kusatol, rasikal, sodium chlorate, tumbleleaf, 500g, 98% min. (Iodometry), CHEBI:65242, Yellow, 231-889-5, 161.97, GHS Signal Word: Danger

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Oxidation reagent
Color Yellow
Melting Point 792.0°C
Physical Form Crystal, Crystalline Powder, Crystals or Crystalline Powder
Quantity 500g
Assay Percent Range 98% min. (Iodometry)
Linear Formula Na2CrO4
Merck Index 15, 8736
Solubility Information Solubility in water: 530g/L (20°C). Other solubilities: soluble in methanol,slightly soluble in ethanol
Formula Weight 161.97
Percent Purity 98+%
Packaging Plastic bottle
Solubility Solubility in water: 530g/L (20°C). Other solubilities: soluble in methanol, slightly soluble in ethanol
Chemical Name or Material Sodium chromate, Anhydrous
Show More Show Less

GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing diffi

GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

EINECSNumber : 231-889-5

RUO – Research Use Only