Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium chromate, 98+%, anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Melting Point | 792.0°C |
Color | Yellow |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 25 g |
Assay Percent Range | 98% min. (Iodometry) |
Linear Formula | Na2CrO4 |
Merck Index | 15, 8736 |
Solubility Information | Solubility in water: 530g/L (20°C). Other solubilities: soluble in methanol, slightly soluble in ethanol |
IUPAC Name | sodium chromate |
Formula Weight | 161.97 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Toxic if swallowed. Harmful in contact with skin. Causes severe skin burns and eye damage. May cause an allergic skin reaction. Fatal if inhaled. May cause allergy or asthma symptoms or breathing diffi |
Precautionary Statement | GHS P Statement Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. |
EINECSNumber | 231-889-5 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction