Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium chromate, anhydrous, 98%

Manufacturer:  Alfa Aesar™ A1054722

Quantity 100g

 View more versions of this product

Catalog No. AAA1054722


  Check Availability
Add to cart

Specifications

Description

Applications
Sodium chromate anhydrous is used as a corrosion inhibitor in the petroleum industry, wood preservative and dyeing auxiliary in the textile industry. It is also used to prepare pigments for paints and inks. It also finds use in pharmaceuticals to determine red blood cell volume. In synthetic chemistry, it is utilized as an oxidant to convert primary alcohols and secondary alcohols to carboxylic acids and ketones respectively.

Solubility
Soluble in water. Slightly soluble in methanol and ethanol.

Notes
Hygroscopic. Incompatible with combustible materials and reducing agents.
Description & Specifications

Chemical Identifiers

7775-11-3
161.97
UN3288
Sodium chromate, Anhydrous
Na2CrO4
MFCD00003484
14,8601

Specifications

7775-11-3
161.97
UN3288
Odorless
100g
98%
Na2CrO4
MFCD00003484
14,8601
792°C
Sodium chromate, Anhydrous

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H330-H334-H340-H350-H360-H372-H314-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : GB2955000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit