Alfa Aesar™ Sodium chromate, anhydrous, 98%

Manufacturer: Alfa Aesar™ A1054736

Quantity 500g

 View more versions of this product

Catalog No. AAA1054736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7775-11-3
    Molecular Formula Na2CrO4
    Formula Weight 161.97
    MDL Number MFCD00003484
    UN Number UN3288
    Merck Index 14,8601
    Chemical Name or Material Sodium chromate, Anhydrous

    Specifications

    Formula Weight 161.97
    Health Hazard 2 GHS H Statement
    H301-H311-H330-H334-H340-H350-H360-H372-H314-H318-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    TSCA TSCA
    CAS 7775-11-3
    Merck Index 14,8601
    Melting Point 792°C
    Quantity 500g
    RTECS Number GB2955000
    Odor Odorless
    UN Number UN3288
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Molecular Formula Na2CrO4
    MDL Number MFCD00003484
    Chemical Name or Material Sodium chromate, Anhydrous
    Assay 98%

    Description

    Applications
    Sodium chromate anhydrous is used as a corrosion inhibitor in the petroleum industry, wood preservative and dyeing auxiliary in the textile industry. It is also used to prepare pigments for paints and inks. It also finds use in pharmaceuticals to determine red blood cell volume. In synthetic chemistry, it is utilized as an oxidant to convert primary alcohols and secondary alcohols to carboxylic acids and ketones respectively.

    Solubility
    Soluble in water. Slightly soluble in methanol and ethanol.

    Notes
    Hygroscopic. Incompatible with combustible materials and reducing agents.

    GHS H Statement
    H301-H311-H330-H334-H340-H350-H360-H372-H314-H318-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit