Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate, Anhydrous, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
68-04-2
C6H5Na3O7
258.07
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
trisodium 2-hydroxypropane-1,2,3-tricarboxylate
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
68-04-2
C6H5Na3O7
258.07
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
trisodium 2-hydroxypropane-1,2,3-tricarboxylate
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

S1986, 68-04-2, C6H5Na3O7

Spectrum™ Sodium Citrate, Anhydrous, USP is used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Percent Purity 99 to 100.5%
Grade USP
Packaging Poly Pail
Water 0.01
Quantity 12 kg