Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate Dihydrate (Granular/Certified), Fisher Chemical™

Click to view available options
Quantity:
500g
3kg
10kg
50kg
275 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6132-04-3
C6H9Na3O9
294.10
MFCD00150031
NLJMYIDDQXHKNR-UHFFFAOYSA-K
trisodium citrate dihydrate,sodium citrate dihydrate,sodium citrate tribasic dihydrate,natriumcitrat-dihydrat,sodium citrate hydrate,1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate,citronensaeure-tri-na-salz-dihydrat,citric acid trisodium salt dihydrate,citric acid, trisodium salt dihydrate,trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
71474
CHEBI:32142
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
6132-04-3
C6H9Na3O9
294.10
MFCD00150031
NLJMYIDDQXHKNR-UHFFFAOYSA-K
trisodium citrate dihydrate,sodium citrate dihydrate,sodium citrate tribasic dihydrate,natriumcitrat-dihydrat,sodium citrate hydrate,1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate,citronensaeure-tri-na-salz-dihydrat,citric acid trisodium salt dihydrate,citric acid, trisodium salt dihydrate,trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
71474
CHEBI:32142
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
Identification Pass Test
Melting Point 300°C
Quantity 275 lb.
Packaging Fiber Drum
Decomposition Information >230°C
Color White
pH 7.5 to 9.5
Solubility Information 0.005% max. Insoluble Matter
Formula Weight 294.1
Physical Form Solid
Chemical Name or Material Sodium Citrate Dihydrate
Show More Show Less

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0