Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate, Dihydrate, Granular, FCC, 99-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6132-04-3
C6H9Na3O9
294.10
NLJMYIDDQXHKNR-UHFFFAOYSA-K
trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
6132-04-3
C6H9Na3O9
294.10
NLJMYIDDQXHKNR-UHFFFAOYSA-K
trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

S1256, 6132-04-3, C6H5Na3O7•2H2O

Spectrum™ Sodium Citrate, Dihydrate, Granular, FCC is used as a flavorant and preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade FCC
CAS Max % 1
Percent Purity 99 to 100.5%
Lead (Pb) 2 mg/kg
Packaging Poly Pail
Water 10-13%
Quantity 12 kg