Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate, Dihydrate, Granular, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
6132-04-3
C6H9Na3O9
294.10
NLJMYIDDQXHKNR-UHFFFAOYSA-K
trisodium citrate dihydrate,sodium citrate dihydrate,sodium citrate tribasic dihydrate,natriumcitrat-dihydrat,sodium citrate hydrate,1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate,citronensaeure-tri-na-salz-dihydrat,citric acid trisodium salt dihydrate,citric acid, trisodium salt dihydrate,trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
71474
CHEBI:32142
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
6132-04-3
C6H9Na3O9
294.10
NLJMYIDDQXHKNR-UHFFFAOYSA-K
trisodium citrate dihydrate,sodium citrate dihydrate,sodium citrate tribasic dihydrate,natriumcitrat-dihydrat,sodium citrate hydrate,1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate,citronensaeure-tri-na-salz-dihydrat,citric acid trisodium salt dihydrate,citric acid, trisodium salt dihydrate,trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
71474
CHEBI:32142
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
Formula Weight 294.1
Physical Form Granular
Grade Multi-Compendial/USP
Packaging Poly Bottle
Specific Gravity 1L = 1.70kg
Quantity 2.5kg
Chemical Name or Material Sodium Citrate, Dihydrate