Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate, Dihydrate, Granular, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

6132-04-3
C6H9Na3O9
294.10
NLJMYIDDQXHKNR-UHFFFAOYSA-K
71474
CHEBI:32142
CAS 6132-04-3
Molecular Formula C6H9Na3O9
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
Molecular Weight (g/mol) 294.10
PubChem CID 71474
ChEBI CHEBI:32142
Formula Weight 294.1
Physical Form Granular
Packaging Poly Bottle
Specific Gravity 1L = 1.70kg
Quantity 2.5kg
Chemical Name or Material Sodium Citrate, Dihydrate
Grade FCC/USP
Show More Show Less