Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate, Dihydrate, Granular, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Manufacturer:  J.T.Baker 409304

Packaging Poly Bottle
Quantity 1kg

 View more versions of this product

Catalog No. 02-004-060

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

    For Liquid Chromatography and Molecular Biology Applications
Description & Specifications

Chemical Identifiers

6132-04-3
294.1
Ultrapure Bioreagent
C6H5Na3O7·2H2O
Sodium Citrate, Dihydrate

Specifications

6132-04-3
C6H5Na3O7·2H2O
Poly Bottle
1kg
Ultrapure Bioreagent
Granular
294.1
1L = 1.70kg
Sodium Citrate, Dihydrate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit