Sodium Citrate, Dihydrate, Granular, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Manufacturer: J.T.Baker 409300

Packaging Poly Bottle
Quantity 100g

 View more versions of this product

Catalog No. 02-004-057

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 6132-04-3
    Molecular Formula C6H5Na3O7·2H2O
    Formula Weight 294.1
    Chemical Name or Material Sodium Citrate, Dihydrate
    Grade Ultrapure Bioreagent

    Specifications

    Formula Weight 294.1
    Physical Form Granular
    Specific Gravity 1L = 1.70kg
    Packaging Poly Bottle
    Molecular Formula C6H5Na3O7·2H2O
    CAS 6132-04-3
    Quantity 100g
    Chemical Name or Material Sodium Citrate, Dihydrate
    Grade Ultrapure Bioreagent

    Description

      For Liquid Chromatography and Molecular Biology Applications

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit